Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Primair™ PA800 Series Welding Helmet

Catalog No. 19085558
Change view
Click to view available options
9 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-085-558 9 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-085-558 Supplier Honeywell™ Supplier No. PA810

Honeywell Primair™ PA800 Series Welding Helmet features an auto-darkening filter with a light state of 2.5

  • Ergonomic design
  • External controls enable quick transition from welding to grinding, even with gloves on
  • The suspension, helmet and air-duct can all be adjusted to ensure a comfortable fit
  • Lightweight design minimizes head and neck strain and reduces risks of fatigue or injury
  • Cooling comfort
  • Integrated and adjustable air-duct connects easily toto the PA700 PAPR system, to help keep workers cool, comfortable and productive.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance NIOSH 42 CFR 84
Color Black
For Use With (Equipment) PA700 PAPR
Product Type Helmet
Size Welding
Weight (English) 1.5 lb.
Head Style Welding
Width (English) 9 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.