Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Monkey CD31/PECAM-1 ELISA Kit (Colorimetric)
SDP

Catalog No. NBP192720 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP192720 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP192720 Supplier Novus Biologicals™ Supplier No. NBP192720
Only null left
Add to Cart
Add to Cart

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Primate CD31/PECAM-1 ELISA Kit (Colorimetric) measures primate CD31/PECAM-1 in cell culture supernatant, serum (baboon).

Specifications

Assay Range 0.48 - 30.0 U/mL
Assay Sensitivity 0.06 U/mL
Product Type ELISA Kit
Sample Type Baboon Serum, Cell Culture Supernatant
Target Species Primate
For Use With (Application) ELISA
Gene ID (Entrez) 5175
Gene Symbol PECAM1
Kit Contents Sample Diluent, (buffered protein matrix), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films, Aluminum pouch(es) with a Microwell Plate coated with Monoclonal Antibody (murine) to monkey sPECAM-1, HRP-Conjugate anti-monkey-sPECAM-1 monoclonal (murine) antibody, Monkey sPECAM-1 Standard, Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Quantity 96 Tests
Research Discipline Angiogenesis, Cancer, CD Markers, Cellular Markers, Cytoskeleton Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Signal
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule
Target CD31/PECAM-1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.