Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Monkey CXCL8/IL-8 ELISA Kit (Colorimetric)
SDP

Catalog No. NBP192706 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP192706 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP192706 Supplier Novus Biologicals™ Supplier No. NBP192706
Only null left
Add to Cart
Add to Cart

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 

The Primate CXCL8/IL-8 ELISA Kit (Colorimetric) measures primate CXCL8/IL-8 in cell culture supernatant, serum (baboon, cynomolgus).

Specifications

Assay Range 19.5 - 1,250 U/mL
Assay Sensitivity 3.0 U/mL
Product Type ELISA Kit
Sample Type Baboon Serum, Cell Culture Supernatant, Cynomolgus Serum
Target Species Primate
For Use With (Application) ELISA
Gene ID (Entrez) 3576
Gene Symbol CXCL8
Kit Contents Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey IL-8, Biotin-Conjugate anti-monkey IL-8 polyclonal antibody, Streptavidin-HRP, Monkey IL-8 Standard Concentrate, 125000U/mL, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
Quantity 96 Tests
Research Discipline Angiogenesis, Cancer, Cytokine Research, Immunology, Innate Immunity, Signal Transduction
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8SCYB8, Emoctakin, GCP-1TSG-1, interleukin 8, K60, LECT, MDNCFb-ENAP, member 8, MONAPGCP1, NAP-1NAP1, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor
Target CXCL8/IL-8
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.