Learn More
Description
- The Primate IFN-gamma ELISpot Kit is designed for the detection and enumeration of individual cells secreting primate IFN-gamma.
- Quantitative sandwich ELISpot assay provides high sensitivity, enabling the measurement of response frequencies below 1 in 100,000 cells without the need for in vitro cell expansion.
- Assay Type: Quantitative Sandwich ELISA
- Sample Type: Whole Cells
- Format: 96-well PVDF-backed microplate
- Assay Length: 3 hour 15 min. to 4 hour 30 min. (plus plate coating)
- Key Components: Pre-coated PVDF microplate, biotinylated detection antibody, streptavidin-AP conjugate, and positive control
- Validated Species: Primate
Specifications
Specifications
| Conjugate | AP |
| Cross Reactivity | Rhesus Monkey |
| Quantity | 1 Kit |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Product Line | R&D Systems |
| Product Type | ELISpot Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
