Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Monkey IL-10 ELISA Kit (Colorimetric)
SDP

Catalog No. NBP192708 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP192708 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP192708 Supplier Novus Biologicals™ Supplier No. NBP192708
Add to Cart
Add to Cart

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 

The Monkey IL-10 ELISA Kit (Colorimetric) measures monkey IL-10 in cell culture supernatant, serum (baboon, cynomolgus).

Specifications

Assay Range 3.9 - 250 U/mL
Assay Sensitivity 1.3 U/mL
Product Type ELISA Kit
Sample Type Baboon Serum, Cell Culture Supernatant, Cynomolgus Serum
Specificity The interference of circulating factors of the immune systemwas evaluated by spiking these proteins at physiologically relevant concentrations into a monkey IL-10 positive serum. There was no crossreactivity detected.
Target Species Primate
For Use With (Application) ELISA
Gene ID (Entrez) 3586
Gene Symbol IL10
Kit Contents Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey IL-10, Biotin-Conjugate anti-monkey IL-10 monoclonal antibody, Streptavidin-HRP, Monkey IL-10 Standard lyophilized, 500U/mL upon reconstitution, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
Quantity 96 Tests
Research Discipline Adaptive Immunity, Apoptosis, Asthma, Cancer, Cytokine Research, Immunology, Innate Immunity
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym CSIFMGC126450, Cytokine synthesis inhibitory factor, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF
Target IL-10
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.