Learn More
Description
Specifications
Specifications
| Assay Range | 20.0 - 1,250 U/mL |
| Assay Sensitivity | 10.0 U/mL |
| Product Type | ELISA Kit |
| Sample Type | Baboon Serum, Cell Culture Supernatant, Cynomolgus Serum |
| Specificity | The cross reactivity and interference of circulating factors of the immune system was evaluated by spiking these proteins at physiologically relevant concentrations into a monkey IL-2 positive serum. There was no cross reactivity or interference detected. |
| Target Species | Primate |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3558 |
| Gene Symbol | IL2 |
| Quantity | 96 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
