Learn More
Prime Wire & Cable Arctic Blue™ All-Weather Extension Cords
Catalog No. 19817286
Change view
Click to view available options
100 ft. long
25 ft. long
50 ft.long
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-817-286 25 ft. long
19-817-287 50 ft.long
19-817-288 100 ft. long
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-817-286 Supplier Prime Wire & Cable Supplier No. EC630825
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Extra heavy-duty Prime-Flex-2™ TPE-rubber triple tap extension cords designed for use by contractors and industrial personnel in extreme temperatures—from -58 to +221°F.
  • Resist water, ozone, oils, chemicals and rough handling under the most severe operating conditions and are flame retardant
  • Distinctive orange stripe extruded into blue cord and matching orange connectors to enhance visibility.
  • Will not stiffen or crack even in the most extreme weather conditions
  • Provide multi-outlet service centers where needed
  • Three-conductor SJEOW, 300V 15A, 1875 watts, 12/3 AWG. UL and CUL listed.
Compliance
OSHA compliant.