Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Primuline, TCI America™
SDP

Catalog No. P06035G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P06035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P06035G Supplier TCI America Supplier No. P06035G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 8064-60-6
Molecular Formula C21H14N3NaO3S3
Molecular Weight (g/mol) 475.531
MDL Number MFCD00005798
InChI Key RSRNHSYYBLEMOI-UHFFFAOYSA-M
Synonym primuline, primulin, 2,6'-bibenzothiazole-7-sulfonic acid, 2'-4-aminophenyl-6-methyl-, monosodium salt, c.i. direct yellow 59, monosodium salt, sodium 2'-4-aminophenyl-6-methyl 2,6'-bibenzothiazole-7-sulphonate, 2,6'-bibenzothiazole-7-sulfonic acid, 2'-4-aminophenyl-6-methyl-, sodium salt 1:1, c.i. direct yellow 59, monosodium salt 8ci, primuline, dye content 50 %
PubChem CID 3769888
IUPAC Name sodium;2-[2-(4-aminophenyl)-1,3-benzothiazol-6-yl]-6-methyl-1,3-benzothiazole-7-sulfonate
SMILES CC1=C(C2=C(C=C1)N=C(S2)C3=CC4=C(C=C3)N=C(S4)C5=CC=C(C=C5)N)S(=O)(=O)[O-].[Na+]

Specifications

Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Primuline
RTECSNumber TV1050000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.