Specifications
| Pore Style | Small Square Pores |
| Quantity | 4 x 250 cassettes (1000)/cs |
| Closure Included | Separate lids included |
| Closure Type | Reversible |
| PrintMate Compatible | Yes |
| Color | Light Green |
| Product Type | PrintMate Biopsy Cassette for PrintMate Printers |