Learn More
Epredia™ PrintMate™ Biopsy Cassettes
Catalog No. B851110LI Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Quantity:
10 x 75 cassettes (750)/cs
4 x 250 cassettes (1000)/cs
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
B851110LI Lilac 10 x 75 cassettes (750)/cs
B851080BE Beige 4 x 250 cassettes (1000)/cs
B851080BL Blue 4 x 250 cassettes (1000)/cs
B851080GN Green 4 x 250 cassettes (1000)/cs
B851080GY Gray 4 x 250 cassettes (1000)/cs
B851080LA Lavender 4 x 250 cassettes (1000)/cs
B851080LG Light Green 4 x 250 cassettes (1000)/cs
B851080LI Lilac 4 x 250 cassettes (1000)/cs
B851080OR Orange 4 x 250 cassettes (1000)/cs
B851080PK Pink 4 x 250 cassettes (1000)/cs
B851080TN Tan 4 x 250 cassettes (1000)/cs
B851080WH White 4 x 250 cassettes (1000)/cs
B851080YW Yellow 4 x 250 cassettes (1000)/cs
B851110BE Beige 10 x 75 cassettes (750)/cs
B851110BL Blue 10 x 75 cassettes (750)/cs
B851110GN Green 10 x 75 cassettes (750)/cs
B851110GY Gray 10 x 75 cassettes (750)/cs
B851110LA Lavender 10 x 75 cassettes (750)/cs
B851110LG Light Green 10 x 75 cassettes (750)/cs
B851110OR Orange 10 x 75 cassettes (750)/cs
B851110PK Pink 10 x 75 cassettes (750)/cs
B851110TN Tan 10 x 75 cassettes (750)/cs
B851110WH White 10 x 75 cassettes (750)/cs
B851110YW Yellow 10 x 75 cassettes (750)/cs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
Catalog No. B851110LI Supplier Epredia™ Supplier No. B851110LI
Add to Cart
Add to Cart
Epredia™ PrintMate Cassettes™ are designed to contain incised tissue safely and securely for tissue processing, embedding and sectioning. PrintMate Cassettes provide exceptional scanning accuracy and reliability when used with the Epredia™ PrintMate™ and Epredia™ SlideMate™ barcode scanner.
Designed for performance and user convenience, the PrintMate cassette range has been optimized for superior workflow and printing with the PrintMate Classic and PrintMate AS printing systems. The even, textured printing face provides the optimal surface for high-quality printing every time. For additional information, the right and left sides are textured for clear handwriting with suitable pens. The lid catches have been adjusted for ease of use with low force opening and closing, while still giving the security needed while processing.Available in tubes for automatic loading or loose for manual use, and in twelve colors for enhanced workflow identification.
Specifications
Pore Style Small Square Pores
Quantity 10 x 75 cassettes (750)/cs
Closure Included Separate lids included
Closure Type Reversible
PrintMate Compatible Yes
Color Lilac
Product Type PrintMate Biopsy Cassette for PrintMate Printers
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov