Description
- For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
- Reduces the number of locks being used
- Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
- Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
| Product Type | Mechanical Risk Lockouts |
| Color | Red |
| Material | Powder-coated Metal |
| Includes | (3) 1.5 in steel lockout hasp; (1) number sheet; (1) osha lockout summary sheet; (3) red padlock; (5) lockout tag with ties; (2) magnetic danger signs; (1) lockout keychain; (1) pad of 50 lockout/tagout checklist forms |
| Quantity | 1 Kit |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.