Learn More
Brady™ Maintenance Station Parts
Catalog No. 19104051 Shop All Brady Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
19-104-051 1 Kit
1 options
Catalog No. 19-104-051 Supplier Brady™ Supplier No. 45250
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Maintenance Station Parts is part of a safety plan to protect workers and the worksite using lockout products. Lockout devices are an important part of safety protocol.
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Quantity 1 Kit