Description
- A simple solution for electric equipment lockout procedures
- Accommodates a large variety of plug shapes and sizes
Specifications
Specifications
| Product Type | Plug Lockout |
| Color | Yellow |
| Material | Thermoplastic Elastomer |
| Dimensions (L x W x H) | 3.47 x 6.25 x 5.2 in. |
| Quantity | 48/Pack |
Safety and Handling
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.