Learn More
Brady™ Valve Lockout Starter Kit
Catalog No. 19104217 Shop All Brady Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
19-104-217 1 Kit
1 options
Catalog No. 19-104-217
Supplier: Brady™ LK095E
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Valve Lockout Starter Kit is part of a safety plan to protect workers and the worksite using lockout products. Lockout devices are an important part of safety protocol.
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Product Type Mechanical Risk Lockouts
Quantity 1 Kit
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.