Learn More
United Scientific™ Prism Microscope
SDPSCI_ED
Catalog No. S43605 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Head Style:
Single Eyepiece
Catalog No. Head Style
S43605 Single Eyepiece
1 options
Catalog No. S43605 Supplier United Scientific™ Supplier No. MCRPR1
Add to Cart
Add to Cart
This Prism Microscope is ideal for classroom or field use.
  • Microscope body, viewing tube and stage are constructed of cast metal.
  • Light-gathering lucite prism provides sub-stage illumination, without need for electricity, by using ambient or room light.
  • Tilt mechanism enables users to adjust angle of viewing tube while slides are held securely in place by spring-loaded stage clips.
  • Dimensions (D x W x H): 5.5 x 4 x 10.5 in.
Specifications
Eyepiece WF 10X
Lighting Type Light Gathering Lucite Prism for Sub Stage Illumination
Head Style Single Eyepiece
Product Type Prism Microscope
Magnification Power 40X
Objective Types Fixed
For Use With (Application) Ideal for Classroom or Field Use