Learn More
TARGETMOL CHEMICALS INC PRITELIVIR 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T250450MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pritelivir (BAY 57-1293) (BAY 57-1293) is a potent helicase-primase inhibitor with active against HSV-1 and HSV-2 (IC50 20 nM). purity: 99%
Catalog No. 50-257-7104