Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ PRKCB1 (Human) Recombinant Protein

Catalog No. 89339445 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-445 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-445 Supplier Abnova™ Supplier No. P6542
Add to Cart
Add to Cart

For use with Functional Study.

Human PRKCB1 (NP_997700.1, 1 a.a. - 671 a.a.) full length recombinant protein with GST-tag at N-terminal using baculovirus expression system.

Specifications

For Use With (Application) Functional Study
Gene ID (Entrez) 5579
Name protein kinase C, beta
Quality Control Testing The purity was assessed by SDS-PAGE/CBB staining.
Quantity 5 μg
Storage Requirements Stored at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias MGC41878, PKC-beta, PKCB, PRKCB1, PRKCB2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.