Learn More
Invitrogen™ Human PRKD1 (PKC mu), GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.
Specifications
Specifications
| Accession Number | O94806, Q15139, Q9BZL6 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50mM tris with 0.02% Triton X-100, 150mM NaCl, 0.5mM EDTA, 50% glycerol, 2mM DTT and no preservative; pH 7.5 |
| Gene ID (Entrez) | 23683, 25865, 5587 |
| Molecular Weight (g/mol) | 129.3 kDa |
| Name | Human PRKD1 (PKC mu), GST Tag |
| pH Range | 7.5 |
| Purification Method | Purified |
| Quantity | 10 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.