Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRKY Monoclonal Antibody (OTI6H3)
GREENER_CHOICE

Catalog No. PIMA524955
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524955 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524955 Supplier Invitrogen™ Supplier No. MA524955
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PRKY Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

A group of proteins that are intimately involved in this process are the serine/ threonine (Ser/Thr) protein kinases. PRKY (protein kinase, Y-linked) and PRKX (protein kinase, X-linked) are members of the Ser/Thr protein kinase family, both of which belong to the subfamily of cAMP-dependent kinases. Encoded by a gene that is located near the pseudoautosomal region on chromosome Y, PRKY contains one protein kinase domain through which it catalyzes the ATP-dependent phosphorylation of target proteins. Functioning in a similar manner to PRXY, PRKX contains one protein kinase domain and is essential for macrophage differentiation and renal epithelial cell migration. Defects in the genes encoding PRKY and PRXX are associated with sex reversal disorder, namely XX in males and XY in females.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRKY
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI6H3
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PRKY
Gene Accession No. O43930
Gene Alias PRKXP3; PRKY; PRKYP; protein kinase, Y-linked, pseudogene
Gene Symbols PRKY
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human PRKY expression vector.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5616
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.