Learn More
Med Vet International Adhesive Dressing 3M Medipore 2-3/8 X 4 inch Soft Cloth Rectangle White Sterile, 3564, 200/Case
Supplier: Med Vet International 315359CS
3M's Medipore +Pad soft cloth dressings are porous and breathable to maintain skin integrity They have a nonadherent pad that won't unsettle the wound bed The pads are absorbent to help maintain a proper healing environment The adhesive on the white dressings is strong yet gentle on the skin These sterile dressings conform around contours and flex with movement to provide optimal protection 2-3/8 x 4 inch adhesive dressing has a 1 x 2-3/4 inch pad
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.