Learn More
PRMT2 Mouse anti-Human, Clone: OTI3B3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503588

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
PRMT is a member of the mammalian PRMT (protein arginine N-methyltransferase) family. The family consists of 7 members that share two large domains of homology. Outside of these domains, epitopes are identified and antibodies against all 7 PRMT members have been developed.These enzymes have been shown to have essential regulatory functions by methylation of key proteins in several fundamental areas. These proteins include nuclear proteins, IL enhancer binding factor, nuclear factors, cell cycle proteins, signal transduction proteins, apoptosis proteins, and viral proteins.Specifications
PRMT2 | |
Monoclonal | |
Unconjugated | |
PRMT2 | |
HRMT1L1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
3275 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI3B3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P55345 | |
PRMT2 | |
Full length human recombit protein of human PRMT2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.