Learn More
Invitrogen™ PRMT4 Monoclonal Antibody (3H2)

Description
MA5-15796 targets CARM1 in indirect ELISA, FACS, IF, IHC, and WB applications and shows reactivity with Human, Non-human primate, Mouse and Rat samples. The MA5-15796 immunogen is purified recombinant fragment of human CARM1 expressed in E. Coli. MA5-15796 detects CARM1 which has a predicted molecular weight of approximately 65kDa.
Specifications
Specifications
| Antigen | PRMT4 |
| Applications | ChIP Assay, ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 3H2 |
| Concentration | Conc. not determined |
| Conjugate | Unconjugated |
| Formulation | ascites with 0.03% sodium azide |
| Gene | Carm1 |
| Gene Accession No. | Q4AE70, Q86X55, Q9WVG6 |
| Gene Alias | Carm1; coactivator associated arginine methyltransferase 1; coactivator-associated arginine methyltransferase 1; coactivator-associated arginine methyltransferase 1 variant 5; histone-arginine methyltransferase CARM1; PRMT4; protein arginine methyltransferase; protein arginine N-methyltransferase 4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.