Learn More
Invitrogen™ PRMT5 Polyclonal Antibody

Description
Suggested positive control: HeLa, NIH-3T3, Hela whole cell extract, antigen standard for PRMT5 (transient overexpression lysate).
Specifications
Specifications
| Antigen | PRMT5 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 100μg/mL BSA and 0.02% sodium azide |
| Gene | PRMT5 |
| Gene Accession No. | O14744, Q8CIG8 |
| Gene Alias | 72 kDa ICln-binding protein; EC 2.1.1.125; histone-arginine N-methyltransferase PRMT5; HMT1 hnRNP methyltransferase-like 5; HRMT1L5; hypothetical protein; I79_013398; IBP72; Jak-binding protein 1; JBP1; PRMT5; protein arginine methyltransferase 5; protein arginine N-methyltransferase 5; Protein arginine N-methyltransferase 5, N-terminally processed; protein arginine N-methyltransferase 5-like; shk1 kinase-binding protein 1 homolog; Skb1; SKB1 homolog; SKB1Hs |
| Gene Symbols | PRMT5 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.