Learn More
Invitrogen™ PRMT6 Polyclonal Antibody

Description
Suggested positive control: HeLa or RAW. Immunogen sequence: CEEDGAEREA ALERPRRTKR ER.
Specifications
Specifications
| Antigen | PRMT6 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | PRMT6 |
| Gene Accession No. | Q6NZB1, Q96LA8 |
| Gene Alias | AW124876; BB233495; FLJ10559; FLJ51477; heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 6; Histone-arginine N-methyltransferase PRMT6; HMT1 hnRNP methyltransferase-like 6; Hrmt1l6; PRMT6; PRMT-6; protein arginine methyltransferase 6; protein arginine N-methyltransferase 6 |
| Gene Symbols | PRMT6 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.