Learn More
Honeywell™ PRO 3003 Leather Station/Duty Boots
Extremely lightweight and comfortable
Supplier:  Honeywell™ BT30030852
 View more versions of this product
Catalog No. 19-168-826
Only null left
Add to Cart
Description
Description
Its oblique toe, Cambrelle lining, and stitched-down sole coupled with Military AB leather create a boot that is the most water-resistant available in the fire service. The unique design makes this the perfect boot for station/duty use.
  • Water resistant leather for durability, breathability and comfort
  • Oblique-shaped steel toe for added protection and comfort
  • Reinforced stitched-down sole
  • Abrasion and bacteria resistant Cambrelle lining
  • Foam-filled comfort collar that will not chafe
  • Dual function closure system for enhanced ankle support and fit plus zipper for rapid donning and doffing
  • Thermoplastic backpart molded heel counter ensures superior comfort, fit and durability; does not lose shape if exposed to moisture
  • Full wraparound Goodyear™ welt adds support, reduces fatigue and ankle sprains/strains
  • Welted construction retains its shape over the life of the boot so you keep that great fit
  • Triple linear ridge shank for increased arch support and puncture resistant steel bottom plate
  • Vibram Technical Lug non-marking outsole, combined with special fire retardant NFPA compound, for improved traction and durability
  • Full wrap-around Goodyear welt adds support and reduces fatigue and ankle strains and sprains
  • Nonmarking Vibram™ lug sole
  • Thermoplastic backpart molded heel counter maintains shape in moist conditions
  • Triple linear ridge shank for increased arch support
  • Puncture-resistant steel bottom plate
  • 8 in. high
This boot does not comply with any NFPA standards
Order Info
D width shown; also available in E and EEE widths.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
8 in.
Cambrelle
Leather
8.5D
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.