Learn More
Honeywell™ PRO 3003 Leather Station/Duty Boots
Extremely lightweight and comfortable
Supplier: Honeywell™ BT30031152
Description
Its oblique toe, Cambrelle lining, and stitched-down sole coupled with Military AB leather create a boot that is the most water-resistant available in the fire service. The unique design makes this the perfect boot for station/duty use.
- Water resistant leather for durability, breathability and comfort
- Oblique-shaped steel toe for added protection and comfort
- Reinforced stitched-down sole
- Abrasion and bacteria resistant Cambrelle lining
- Foam-filled comfort collar that will not chafe
- Dual function closure system for enhanced ankle support and fit plus zipper for rapid donning and doffing
- Thermoplastic backpart molded heel counter ensures superior comfort, fit and durability; does not lose shape if exposed to moisture
- Full wraparound Goodyear™ welt adds support, reduces fatigue and ankle sprains/strains
- Welted construction retains its shape over the life of the boot so you keep that great fit
- Triple linear ridge shank for increased arch support and puncture resistant steel bottom plate
- Vibram Technical Lug non-marking outsole, combined with special fire retardant NFPA compound, for improved traction and durability
- Full wrap-around Goodyear welt adds support and reduces fatigue and ankle strains and sprains
- Nonmarking Vibram™ lug sole
- Thermoplastic backpart molded heel counter maintains shape in moist conditions
- Triple linear ridge shank for increased arch support
- Puncture-resistant steel bottom plate
- 8 in. high
This boot does not comply with any NFPA standards
Order Info
D width shown; also available in E and EEE widths.
Specifications
8 in. | |
Cambrelle | |
Leather | |
11.5D |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.