Learn More
Honeywell™ PRO 5006 Structural Firefighting Boots, D Width
Superior combination of comfort and absorbency
Supplier: Honeywell™ BT5006B1152
Description
Quilted to 7.5 oz Kevlar™ felt thermal barrier and laminated to fully encapsulating Crosstech™ Footwear Fabric bootie. Crosstech™ moisture barrier provides superior liquid penetration resistance to blood, body fluids and NFPA tested common chemicals.
- Military AB leather construction insulates and adds abrasion-, puncture-, water- and chemical-resistance
- With heavy duty 0.75 in. reinforced straps mounted to the exterior of boot and anchored with a 2 × 1.5 in. shield-shaped base
- External texon board shinguard protects and guards sensitive shin area of legs
- Patented PowerToe™ cover resists toe abrasion and provides unparalled wear over the lifetime of the boots
- Oblique toe safety cap provides additional room in height and width of boot
- Fire/Ice PowerSole™ angled lugs provide substantial edging and gripping capabilities
- Molded thermoplastic heel counter locks foot into the boot and anchors it in place
- Patented PowerHeel™ cover protects heel area from abusive doffing of boots
- Corrison-resistant steel ladder shank and stainless steel, full length, toe-to-heel puncture resistant bottom plate
Order Info
D-widths shown in table below. Wide widths also available. Available through Fisher Safety's GSA Advantage!™ Schedule 84.
Compliance
Meet NFPA 1971, 2007 Edition. Berry Amendment compliant.
Specifications
14 in. | |
Leather, Dupont Kevlar Aramid Fiber, CrossTech | |
11.5D |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.