Learn More
Honeywell™ PRO 5007 Structural Firefighting Bunker Boots - E Width
Description
- Exclusive bootie package has a Cambrelle a full-height Crosstech™ moisture barrier and 7.5 oz Kevlar™ thermal barrier, protection for feet and legs
- Texon board shinguard cushions and protects shins when standing on ladders
- Heavy-duty round handle straps prevent chafed legs and protect thermal barrier of bunker pants
- Reinforced side and back stays heavy-duty waterproof leather ensures the boots will remain upright and retain their shape with traditional bunker pant stacking
- Breathable Advance™ ripstop fabric significantly reduces the weight of the boot while maintaining durability and flexibility
- Supportive Goodyear welt construction allows flow through of air in the sole area, keeping the sole area ventilated, durable, and strong. Molds to the wearer's foot for extreme long-term comfort
- Premium quality military AB leather full gain, flexible, durable, and water resistant
- Reflective welt is visible below the pant line, aiding firefighter visibility
- PowerToe protection improves life cycle of the toe area by 5 times
- Broad oblique steel toe fits even the widest of feet and offers extreme comfort
- Vibram™ Lug broad-base sole offers improved stability over profiled soles. Lug pattern of traditional stars provides maximum wear, traction, and durability, all with improved structural balance to the wearer
- Secure heel system is strategically placed to substantially reduce heel slippage
- Visible PowerHeel™ protects the heel area from damage as a result of improper doffing of boots
- Removable molded insert
- Corrosion-resistant triple rung shank for superb arch and ladder support
- Puncture resist stainless steel bottom plate exceeds NFPA standard for incidental punctures on the fire ground Widths: E Full sizes: 5 to16; Half sizes: 5 1/2 to 12 1/2
Compliance
NFPA 1971 2007 edition and Class 75 impact resistance/compression resistance requirements of ASTM F2413-05
Specifications
Specifications
| Height (English) | 14 in. |
| Size | 10E |