Learn More
CPC Scientific H-Glu-Leu-Arg-Met-Ser-Ser-Ser-Tyr-Pro-Thr-Gly-Leu-Ala-Asp-Val-Lys-Ala-Gly-Pro-Ala-Gln-Thr-Leu-Ile-Arg-Pro-Gln-Asp-Met-Lys-Gly-Ala-Ser-Arg-Ser-Pro-Glu-Asp-Ser-Ser-Pro-Asp-Ala-Ala-Arg-Ile-Arg-Val-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ADRM008B
Encompass
The adrenomedullin (ADM) precursor fragment is used in diagnosing several disease states, such as the dysfunction of the cardiovascular system and sepsis. The released amounts of these peptides is a reflection of the stoichiometric generation of ADM and proADM. ADM cannot be easily determined directly, because of the masking by binding proteins, instability in serum and stickiness to surfaces. ONE-LETTER SEQUENCE: ELRMSSSYPTGLADVKAGPAQTLIRPQDMKGASRSPEDSSPDAARIRVMOLECULAR FORMULA: C215H359N67O73S2MOLECULAR WEIGHT:5114.8STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [166798-69-2], RESEARCH AREA: CardiovascularREFERENCES: J.Struck et al., Peptides , 25, 1369 (2004)
Catalog No. 50-210-8630
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.