Learn More
CPC Scientific H-Ala-Arg-Leu-Asp-Val-Ala-Ala-Glu-Phe-Arg-Lys-Lys-Trp-Asn-Lys-Trp-Ala-Leu-Ser-Arg-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ADRM013B
Encompass
Prodepin, porcine.ONE-LETTER SEQUENCE: ARLDVAAEFRKKWNKWALSR-NH2MOLECULAR FORMULA: C112H178N36O26MOLECULAR WEIGHT:2444.9STORAGE CONDITIONS: -20 5CRESEARCH AREA: Cardiovascular
Catalog No. 50-210-8640
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.