Learn More
ABclonal Technology Pro-Collagen I/COL1A1
SDP
Supplier:  ABclonal Technology A26946100UL
Encompass_Preferred
This gene encodes the pro-alpha1 chains of type I collagen whose triple helix comprises two alpha1 chains and one alpha2 chain Type I is a fibril-forming collagen found in most connective tissues and is abundant in bone cornea dermis and tendon Mutations in this gene are associated with osteogenesis imperfecta types I-IV Ehlers-Danlos syndrome type VIIA Ehlers-Danlos syndrome Classical type Caffey Disease and idiopathic osteoporosis Reciprocal translocations between chromosomes 17 and 22 where this gene and the gene for platelet-derived growth factor beta are located are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans resulting from unregulated expression of the growth factor Two transcripts resulting from the use of alternate polyadenylation signals have been identified for this gene
Catalog No. 50-000-12467
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.