Learn More
Pro-Lab Diagnostics™ AmnioTest™ Kits
SDP
Catalog No. 23295691 Shop All Pro-Lab Diagnostics Products
Change view
Click to view available options
Quantity:
100 Pack
20 Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-295691 100 Pack
23-295-696 20 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-295691 Supplier Pro-Lab Diagnostics™ Supplier No. PL.901
Add to Cart
Add to Cart
This item is not returnable. View return policy
Assists in the detection of fetal membrane rupture
  • Offers an efficient, cost-effective, reliable, and simple-to-use point-of-care screen test to detect amniotic fluid in near-term patients
  • An invaluable tool for OB-GYN and L+D clinics
  • Sterilized by gamma irradiation
  • Packaged individually in peel-pouches, one hundred per box
  • Refill buffers and color interpretation cards available, see 22-286-164 and 22-286-158
Specifications
Type Amniotic Fluid Test Kit
For Use With (Application) Detection of fetal membrane rupture and also an invaluable tool for OB-GYN and L+D clinics
Includes Disposable swabs impregnated with Nitrazine yellow dye packed in individual sterilized sleeve, AmnioTest™ colour card (PL.902) and product instructions for use
Quantity 100 Pack