Learn More
Pro-Lab Diagnostics™ Pro-Loops™ Insulated Inoculating Loop Wire Holders
SDP
Catalog No. 22286243 Shop All Pro-Lab Diagnostics Products
Change view
Click to view available options
Product Type:
Inoculating Loop Holder
Insulated Loop Holder
Length (Metric):
15.2 cm
20.3 cm
Material:
Lightweight Aluminum
Nickel-plated Brass
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Length (Metric) Material
22-286-243 Insulated Loop Holder 15.2 cm Nickel-plated Brass
22-286-244 Inoculating Loop Holder 20.3 cm Nickel-plated Brass
22-286-174 Inoculating Loop Holder 20.3 cm Lightweight Aluminum
22-286-145 Inoculating Loop Holder 15.2 cm Lightweight Aluminum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22-286-243 Supplier Pro-Lab Diagnostics™ Supplier No. PL.106/61
Add to Cart
Add to Cart
Available in nickel-plated brass or lightweight aluminum
  • Individual loop
  • Constructed of nickel-plated brass or aluminum
Specifications
Material Nickel-plated Brass
Length (English) 6 in.
Length (Metric) 15.2 cm
Product Type Insulated Loop Holder
For Use With (Equipment) Pro-Loops™ inoculating loops