Learn More
Description
Includes
- Staph Xtra Latex Reagent: 2 x 2.5mL (PL.1080), 2 x 7.5mL (PL.1081)
- Negative Control Latex Reagent: 5.0mL (PL.1080), 2 x 7.5mL (PL.1081)
- Agglutination Cards: 20 (PL.1080), 48 (PL.1081)
- Mixing Sticks: 200 (PL.1080), 300 (PL.1081)
- Rapid clear results within 20 seconds
- Uses blue latex particles (CMMP) against a white background
Specifications
Specifications
| Detectable Analytes | S. aureus |
| Type | Staph Xtra Latex Kit |
| Includes | 2 x 2.7mL Staph Xtra Latex reagent, 7.5mL negative control Latex, 48 Agglutination cards |
| Quantity | 300 Tests |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
