Learn More
Pro-Lab Diagnostics™ Prolex™ Individual Extraction Reagents and Extraction Set
Shop All Pro-Lab Diagnostics Products

Description
- Instant results obtained at room temperature
- Advanced extraction process using modified nitrous acid reagents
- As few as one colony required
- Colour coded reagents and vials for ease of selection
Specifications
Specifications
| Color Code | White |
| Content And Storage | 2°C to 8°C |
| Type | Streptococcal Reagent Set |
| Includes | 8mL Extraction R1, 8mL extraction R2, 8mL extraction R3 |
| Quantity | 150 Tests |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.