Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pro-Lab Diagnostics™ Prolex™ Individual Extraction Reagents and Extraction Set
SDP

Catalog No. 22286185 Shop All Pro-Lab Diagnostics Products
Change view
Click to view available options
Quantity:
150 Tests
2 x 8 mL
3.2 mL
Type:
Extraction R1
Extraction R2
Extraction R3
Streptococcal Reagent Set
Color Code:
Blue
Red
White
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type Color Code
22-286-185 3.2 mL Extraction R1 Yellow
22-286-187 2 x 8 mL Extraction R3 Blue
22-286-186 3.2 mL Extraction R2 Red
22-286-177 150 Tests Streptococcal Reagent Set White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22-286-185 Supplier Pro-Lab Diagnostics™ Supplier No. PL.037
Add to Cart
Add to Cart

Identifies all major streptococcal groups including Group D using Prolex™ Streptococcal Grouping Latex Kit

  • Instant results obtained at room temperature
  • Advanced extraction process using modified nitrous acid reagents
  • As few as one colony required
  • Colour coded reagents and vials for ease of selection

Specifications

Color Code Yellow
Content And Storage 2°C to 8°C
Detectable Analytes Streptococci (Group A, B, C, D, F, G)
Type Extraction R1
Includes 3.2mL reagent with 0.098% Sodium Azide
Quantity 3.2 mL
Test Time 1 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.