Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pro-Lab Diagnostics™ Prolex™ Streptococcal Grouping Latex Kits
SDP

Catalog No. 22286179 Shop All Pro-Lab Diagnostics Products
Change view
Click to view available options
Quantity:
2 mL
3 mL
Type:
Group A Latex Reagent
Group B Latex Reagent
Group C Latex Reagent
Group D Latex Reagent
Group F Latex Reagent
Group G Latex Reagent
Polyvalent Positive Control
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
22-286-179 3 mL Group B Latex Reagent
22-286-178 3 mL Group A Latex Reagent
22-286-180 3 mL Group C Latex Reagent
22-286-183 3 mL Group F Latex Reagent
22-286-181 3 mL Group D Latex Reagent
22-286-184 3 mL Group G Latex Reagent
22-286-188 2 mL Polyvalent Positive Control
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 22-286-179 Supplier Pro-Lab Diagnostics™ Supplier No. PL.032
Add to Cart
Add to Cart
This item is not returnable. View return policy

For grouping Lancefield betahaemolytic Streptococci

  • Instant results obtained at room temperature
  • Advanced extraction process using modified nitrous acid reagents
  • Enhanced results with blue carboxylate-modified microparticles (CM-MP)
  • As few as one colony required
  • Colour coded reagents and vials for ease of selection
  • Available individually

Specifications

Content And Storage 2°C to 8°C
Type Group B Latex Reagent
Includes 3mL Blue Latex particles coated with purified Rabbit Antibodies
Quantity 3 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.