Learn More
Pro-Lab Diagnostics™ Prolex™ Streptococcal Grouping Latex Kits
Shop All Pro-Lab Diagnostics Products

Description
- Instant results obtained at room temperature
- Advanced extraction process using modified nitrous acid reagents
- Enhanced results with blue carboxylate-modified microparticles (CM-MP)
- As few as one colony required
- Colour coded reagents and vials for ease of selection
- Available individually
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Type | Group D Latex Reagent |
| Includes | 3mL Blue Latex particles coated with purified Rabbit Antibodies |
| Quantity | 3 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.