Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pro-Lab Diagnostics™ TestOxidase™ Reagent
SDP

Catalog No. 22286170 Shop All Pro-Lab Diagnostics Products
Change view
Click to view available options
Quantity:
15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-286-170 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-286-170 Supplier Pro-Lab Diagnostics™ Supplier No. PL.390
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For determination of bacterial cytochrome oxidase

  • Ready to use
  • Composition: 0.3% N,N,N',N'-tetramethyl-p-phenylenediamine in 0.003M buffer containing reducing agents and organic stabilisers

Specifications

Certifications/Compliance CE Marked
Content And Storage 15°C to 30°C
Type TestOxidase™ Reagent
Formulation N,N,N',N'- Tetramethyl-p-Phenylenediamine 0.3% in Buffer 0.003M containing reducing agents and organic Stabilizers
For Use With (Application) Qualitative method for the determination of bacterial Cytochrome Oxidase
Quantity 15 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.