Learn More
Abcam ProANP Peptide, Synthetic, Human, >95% Purity, 100 μg
Description
- Peptide Type: Synthetic
- Species Reactivity: Human
- Purity: >95%
- Tested Applications: BL
- Form: Lyophilized; Constituents: 0.1% Acetic acid
- Storage Conditions: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | ProANP (Human) |
| Substrate Properties | Peptide-based Substrate |
| Shipping Condition | Blue Ice |
| Reactivity | Human |
| Form | Lyophilized |
| Product Type | Blocker and Diluent |
| Buffer | 0.1% acetic acid |
| Content And Storage | Store at -20°C. |
| Label or Dye | Unconjugated |
| Cross Reactivity | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.