Learn More
Apexbio Technology LLC Fisetin 528-48-3 100mg
SDP
Supplier:  Apexbio Technology LLC N2487100
Encompass_Preferred
Fisetin (CAS 528-48-3) is a polyphenolic compound classified as a natural flavonol It is designed to modulate oxidative stress signaling pathways thereby regulating cellular oxidative balance and neuroprotective processes Fisetin exerts its biological activity primarily through scavenging reactive oxygen species and modulating kinase pathways as well as by inducing apoptosis and inhibiting cell proliferation Additionally it attenuates pro-inflammatory cytokines and modulates neuroprotective signal transduction Based on these pharmacological properties fisetin holds research potential in tumor progression and metastasis neurodegenerative diseases inflammation-mediated disorders and cellular signaling mechanisms
Catalog No. 50-000-96512
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.