Learn More
Enzo Life Sciences Probenecid (5g). CAS: 57-66-9
SDP
Supplier:  Enzo Life Sciences ALX430113G005
Encompass_Preferred
Inhibitor of several ABC-transporters of the subfamily ABCC or MRP (multidrug resistant associated protein). Alternative name: 4-[(Dipropylamino)sulfonyl]benzoic acid. Purity: ≥98% (Titration NaOH 0.1M). Solubility: Soluble in 1M sodium hydroxide. Long Term Storage: Ambient.
Catalog No. 50-200-9206
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.