Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Probenecid, 99.8%, MP Biomedicals™

Catalog No. ICN15637090 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15637090 500 g
ICN15637025 25 g
ICN15637080 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15637090 Supplier MP Biomedicals, Inc Supplier No. 0215637090
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57-66-9
Molecular Formula C13H19NO4S
Molecular Weight (g/mol) 285.36
MDL Number MFCD00038402
InChI Key DBABZHXKTCFAPX-UHFFFAOYSA-N
Synonym probenecid, 4-dipropylsulfamoyl benzoic acid, probenecid acid, probecid, benecid, benemid, benuryl, probalan, probenemid, probexin
PubChem CID 4911
ChEBI CHEBI:8426
IUPAC Name 4-(dipropylsulfamoyl)benzoic acid
SMILES CCCN(CCC)S(=O)(=O)C1=CC=C(C=C1)C(O)=O

Specifications

Color White
Quantity 500 g
Formula Weight 285.4
Percent Purity 0.998
Physical Form Powder
Chemical Name or Material Probenecid
EINECSNumber 200-344-3
Recommended Storage Store at Room Temperature.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.