accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3SOOOHO

CAS RN 57-66-9

IUPAC Name: 4-(dipropylsulfamoyl)benzoic acid
Synonyms: probenecid probenecid acid probenicid probenid idis brand of probenecid probenecid parmed brand valdecasas brand of probenecid 丙磺舒 zenith brand of probenecid probexin
Molecular Weight (g/mol): 285.36
Molecular Formula: C13H19NO4S
InChi Key: DBABZHXKTCFAPX-UHFFFAOYSA-N
SMILES: CCCN(CCC)S(=O)(=O)C1=CC=C(C=C1)C(O)=O
Molecular Weight (g/mol) 
  • (6)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (6)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)