Learn More
Description
Probenecid USP is a white or practically white, fine, crystalline powder. It is practically odorless.
Specifications
Specifications
| Chemical Name or Material | Probenecid, USP |
| Packaging | Jar 300ml, White, HDPE, Round, 53-400, TRIVENI-DB, CT |
| CAS | 57-66-9 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 285.3621 |
| Quantity | 100 g |
| Percent Purity | 98 to 101 % |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
