Learn More
Cytiva ProbeQuant™ G-50 Micro Columns
GSA_VA
Catalog No. 45001487 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
50 Pack
Catalog No. Quantity
45-001-487 50 Pack
1 options
Catalog No. 45-001-487 Supplier Cytiva Supplier No. 28903408
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
For rapid purification of labeled DNA (>20 bases) from unincorporated labeled nucleotides in a volume of 25 to 50μL. Cytiva Life Sciences™ ProbeQuant™ G-50 Micro Columns use spin-column chromatography.
  • Prepacked with Sephadex™ G-50 DNA grade, and designed for use in a microcentrifuge
  • Requires less than four minutes from sample application to collection of purified product
  • A portion of the eluted DNA can be counted in a scintillation counter and compared to an unpurified sample to give an approximation of percent incorporation
  • Tested in nickase, single and double-stranded exonuclease, and RNase assays
Order Info
Conveniently prepacked with Sephadex G-50 DNA Grade and pre-equilibrated in STE containing 0.15% Kathon™ CG/ICP Biocide
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Format Spin Column
Isolation Technology Filtration
For Use With (Application) Rapid purification of labeled DNA (>20 bases) from unincorporated labeled nucleotides in a volume of 25 to 50μL using spin column chromatography
Product Type Column
Quantity 50 Pack