Learn More
Selleck Chemical LLC Probucol S2119-10mM/1mL
SDP
Supplier:  Selleck Chemical LLC S211910MM/1ML
Encompass_Preferred
Probucol (DH-581) is an anti-hyperlipidemic drug by lowering the level of cholesterol in the bloodstream by increasing the rate of LDL catabolism
Catalog No. 50-311-1239
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.