Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Probucol, Thermo Scientific™

Catalog No. AC469490050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469490050 5 g
AC469490250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC469490050 Supplier Thermo Scientific Chemicals Supplier No. 469490050
Add to Cart
This item is not returnable. View return policy

Specifications

Chemical Name or Material Probucol
CAS 23288-49-5
Synonym 4,4 '-(Isopropylidendithio)bis[2,6-di-tert-butylphenol]
Quantity 5 g
Solubility insoluble aqueous solution: soluble in methanol, acetone, acetonitrile, THF
Melting Point 125°C to 128°C
Shipping Condition No special requirements
Product Line Life science
Molecular Formula C31 H48 O2 S2
Physical Form Powder or Crystalline Powder
Color White to Almost-white
Purity ≥97.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.