Learn More
Apexbio Technology LLC BRD4770 1374601-40-7 50mg
SDP
Supplier:  Apexbio Technology LLC B483750
Encompass_Preferred
BRD4770 (CAS 1374601-40-7) is a small-molecule inhibitor targeting the histone methyltransferase G9a with an IC50 of 6 3 M In studies using the PANC-1 pancreatic cancer cell line BRD4770 was shown to decrease levels of di- and trimethylated histone H3 lysine 9 (H3K9) through inhibition of G9a activity This reduction in H3K9 methylation is associated with the induction of cellular senescence and cell death as well as suppression of both adherent-dependent and independent cell proliferation BRD4770 serves as a useful probe for exploring the epigenetic regulation of gene expression and cellular senescence in cancer biology research
Catalog No. 50-000-91126
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.