Learn More
Advanced ImmunoChemical, Inc. PROCALCITONIN, HU RECOMBINANT
Supplier:  Advanced ImmunoChemical, Inc. 8PCALRH0.1 MG
Encompass_Preferred
ANTIGENS/Source:E. coli expression systemDescription:Consists of a polypeptide corresponding to 1-116 amino acid residues of human procalcitonin. Does not contain any tags.Applications:An antigen for immunodetection of human PCTAnalysis:Purity > 95 %Immunoreactivity of recombinant human PCT is checked in sandwich immunoassay with specific monoclonal antibodies.Purification:Affinity and reverse-phase chromatographyPresentation:Lyophilized from 20 mM Tris-HCl, pH 8.0, 150 mM NaCl.Product is recommended to reconstitute with deionized water.Storage Conditions:Before and after reconstitution at -20 °C (between -15 and -30 °C allowed)
Catalog No. NC3566234